o-Tolylisocyanat

Lieferant: Thermo Scientific

Synonyme: Isocyansäure(2-methylphenyl)ester, 2-Methylphenylisocyanat

SB01904.1 SB01904.10 SB01904.50
MAYBSB01904.1EA 0 EUR
MAYBSB01904.1 MAYBSB01904.10 MAYBSB01904.50
o-Tolylisocyanat
o-Tolylisocyanat
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Siedepunkt: 185 °C (1013 hPa)
Dichte: 1,07 g/cm³ (20 °C)
MDL: MFCD00002009
CAS-Nummer: 614-68-6
EINECS: 210-389-0
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR