o-Tolylisothiocyanat

Lieferant: Thermo Scientific
Gefahr

SB01457.10 SB01457.50
MAYBSB01457.10EA 0 EUR
MAYBSB01457.10 MAYBSB01457.50
o-Tolylisothiocyanat
o-Tolylisothiocyanat
Formel: C₈H₇NS
Molecular Weight: 149,22 g/mol
MDL: MFCD00004802
CAS-Nummer: 614-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR