o-Tolylisothiocyanat
Lieferant: FLUOROCHEM
|
Gefahr
|
001280-5G
001280-25G
FLUO001280-5GEA
36.9
EUR
FLUO001280-5G
FLUO001280-25G
o-Tolylisothiocyanat
o-Tolylisothiocyanat
Formel:
C₈H₇NS Molecular Weight: 149,22 g/mol |
MDL:
MFCD00004802 CAS-Nummer: 614-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...