2-Octylphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 2-Octyl isothiocyanate

009555-1G 009555-5G
FLUO009555-1GEA 63.9 EUR
FLUO009555-1G FLUO009555-5G
2-Octylphenylisothiocyanat
2-Octylphenylisothiocyanat
Formel: C₁₅H₂₁NS
Molecular Weight: 247.4 g/mol
MDL: MFCD00060400
CAS-Nummer: 69626-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR