2-Methyl-5-nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 2-Isothiocyanato-1-methyl-4-nitrobenzene

009539-1G 009539-5G
FLUO009539-1GEA 31.1 EUR
FLUO009539-1G FLUO009539-5G
2-Methyl-5-nitrophenylisothiocyanat
2-Methyl-5-nitrophenylisothiocyanat
Formel: C₈H₆N₂O₂S
Molecular Weight: 194,21 g/mol
MDL: MFCD00060549
CAS-Nummer: 86317-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR