2-Isopropylanilin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

268887-100ML 268887-5ML
SIAL268887-100MLEA 113 EUR
SIAL268887-100ML SIAL268887-5ML
2-Isopropylanilin, Sigma-Aldrich®
2-Isopropylanilin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00007720
CAS-Nummer: 643-28-7
EINECS: 211-397-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR