4-Hexylanilin ≥98%

Lieferant: Thermo Scientific
Achtung

H26297.06 H26297.14
H26297.06EA 60.1 EUR
H26297.06 H26297.14
4-Hexylanilin ≥98%
4-Hexylanilin

Purity: 98% CAS No.: 33228-45-4 Molecular Formula: C12H19N Formula Weight: 177.29 Flash Point: 110°(230°F) Refractive Index: 1.5250 Boiling Point: 283-285° MDL No.: MFCD00007927

Formel: C₁₂H₁₉N
Molecular Weight: 177,29 g/mol
Siedepunkt: 283…285 °C
Dichte: 0,919
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007927
CAS-Nummer: 33228-45-4
EINECS: 251-409-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR