p-Toluidin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Methylanilin, 4-Aminotoluol, 4-Toluidin
A13698.0B
A13698.0I
A13698.30
A13698.0BEA
81.8
EUR
A13698.0B
A13698.0I
A13698.30
p-Toluidin ≥99%
p-Toluidin
Purity: 99+%
CAS No.: 106-49-0
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 88°(190°F)
Melting Point: 42-46°
Boiling Point: 199-202°
Beilstein Reference: 471281
Merck Reference: 14,9536
UN No.: 3451
MDL No.: MFCD00007906
Formel:
C₇H₉N Molecular Weight: 107.16 g/mol Siedepunkt: 199…202 °C Schmelzpunkt: 42…46 °C Dichte: 1.05 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007906 CAS-Nummer: 106-49-0 EINECS: 203-403-1 UN: 3451 ADR: 6.1 II Merck Index: 12,09674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...