6-Nitro-o-toluidin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-6-nitroanilin, 6-Nitro-2-methylanilin, 2-Amino-3-nitrotoluol
L01803.14
L01803.22
L01803.14EA
65.6
EUR
L01803.14
L01803.22
6-Nitro-o-toluidin ≥98%
6-Nitro-o-toluidin
Purity: 98%
CAS No.: 570-24-1
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 93-96°
Beilstein Reference: 1868029
UN No.: 2660
MDL No.: MFCD00007744
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Siedepunkt: 120…124 °C (1 mmHg) Schmelzpunkt: 93…96 °C Dichte: 1,19 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007744 CAS-Nummer: 570-24-1 EINECS: 209-329-6 UN: 2660 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...