3-Nitro-p-toluidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Nitro-4-methylanilin, 4-Methyl-3-nitroanilin, 4-Amino-2-nitrotoluol
A12538.14
A12538.22
A12538.14EA
57.1
EUR
A12538.14
A12538.22
3-Nitro-p-toluidin ≥98%
3-Nitro-p-toluidin
Purity: 98%
CAS No.: 119-32-4
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Flash Point: 175°(347°F)
Odour: Characteristic
Beilstein Reference: 638984
UN No.: 2660
MDL No.: MFCD00007910
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.15 g/mol Siedepunkt: 306 Schmelzpunkt: 74…79 °C Dichte: 1.312 Flammpunkt: 175 °C (347 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007910 CAS-Nummer: 119-32-4 EINECS: 204-314-0 UN: 2660 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...