3-Methoxybenzylamin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: m-Anisylamine, 3-(Aminomethyl)anisole

L02054.06 L02054.14
L02054.06EA 46 EUR
L02054.06 L02054.14
3-Methoxybenzylamin ≥98%
3-Methoxybenzylamin

Purity: 98+% CAS No.: 5071-96-5 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 110°(230°F) Refractive Index: 1.5470 Boiling Point: 230-231° Beilstein Reference: 1447182 UN No.: 2735 MDL No.: MFCD00008115

Formel: C₈H₁₁NO
Molecular Weight: 137.18 g/mol
Siedepunkt: 230…231 °C
Dichte: 1.066
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008115
CAS-Nummer: 5071-96-5
EINECS: 225-779-6
UN: 2735

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR