4-Methoxybenzylamin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-(Aminomethyl)anisole, p-Anisylamine
125780250
125781000
125785000
ACRO125780250EA
57.3
EUR
ACRO125780250
ACRO125781000
ACRO125785000
4-Methoxybenzylamin 98%
4-Methoxybenzylamin
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 236…237 °C (760 mmHg) Dichte: 1,05 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008122 CAS-Nummer: 2393-23-9 EINECS: 219-247-2 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5460 to 1.5480 (20°C, 589 nm) |
GC | ≥97.5 % |
Sulfide (S) | ≤0.0002 % |
Carbonate (CO3) | ≤0.001 % |
Phosphate (PO4) | ≤0.00005 % |
Sulfate (SO4) | ≤0.0002 % |
Barium (Ba) | ≤0.00001 % |
Bismuth (Bi) | ≤0.00001 % |
Cadmium (Cd) | ≤0.000005 % |
Calcium (Ca) | ≤0.0001 % |
Chromium (Cr) | ≤0.000005 % |
Cobalt (Co) | ≤0.00001 % |
Copper (Cu) | ≤0.00001 % |
Gallium (Ga) | ≤0.000002 % |
Indium (In) | ≤0.000002 % |
Iron (Fe) | ≤0.00001 % |
Lead (Pb) | ≤0.000005 % |
Lithium (Li) | ≤0.000002 % |
Magnesium (Mg) | ≤0.00005 % |
Manganese (Mn) | ≤0.00001 % |
Molybdenum (Mo) | ≤0.00001 % |
Nickel (Ni) | ≤0.000005 % |
Platinum (Pt) | ≤0.00001 % |
Potassium (K) | ≤0.0001 % |
Sodium (Na) | ≤0.0001 % |
Strontium (Sr) | ≤0.0001 % |
Thallium (Tl) | ≤0.000005 % |
Tin (Sn) | ≤0.00001 % |
Titanium (Ti) | ≤0.00001 % |
Zinc (Zn) | ≤0.00001 % |
Reducing substances | ≤0.0005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...