3-Aminobenzylalkohol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Hydroxymethyl)anilin, (3-Aminophenyl)methanol
184570050
184570250
ACRO184570050EA
33.3
EUR
ACRO184570050
ACRO184570250
3-Aminobenzylalkohol 97%
3-Aminobenzylalkohol
Formel:
C₇H₉NO Molecular Weight: 123.15 g/mol Siedepunkt: 272 Schmelzpunkt: 91…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007817 CAS-Nummer: 1877-77-6 EINECS: 217-514-8 |
Spezifikation Testergebnisse
Appearance | Beige to grey to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 91 to 95 °C |
GC | ≥96.0 % |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.1 % |
Specific optical rotation | +42° to +48° (20 °C, 589 nm) (2 % in 0.1M HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...