3,4-Diaminotoluol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-1,2-phenylendiamin, 1,2-Diamino-4-methylbenzol, Toluol-3,4-diamin, Toluylen-3,4-diamin, 4-Methyl-o-phenylendiamin

B24378.22 B24378.36
B24378.22EA 39 EUR
B24378.22 B24378.36
3,4-Diaminotoluol 97%
3,4-Diaminotoluol

Purity: 97% CAS No.: 496-72-0 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 86-90° Boiling Point: 265° MDL No.: MFCD00007728

Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
Siedepunkt: 265 °C
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007728
CAS-Nummer: 496-72-0
EINECS: 207-826-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR