2,6-Diaminotoluol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Toluol-2,6-diamin, Toluylen-2,6-diamin, 1,3-Diamino-2-methylbenzol, 2-Methyl-m-phenylendiamin

L04278.06 L04278.14
L04278.06EA 34.4 EUR
L04278.06 L04278.14
2,6-Diaminotoluol 97%
2,6-Diaminotoluol

Purity: 97% CAS No.: 823-40-5 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 104-106° Beilstein Reference: 2079476 UN No.: 2811 MDL No.: MFCD00007800

Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
Siedepunkt: 280…282 °C (1013 hPa)
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007800
CAS-Nummer: 823-40-5
EINECS: 212-513-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR