5-Methyl-2-nitroanisol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1-Methoxy-3-methyl-6-nitrobenzol, 3-Methoxy-4-nitrotoluol, Methyl-6-nitro-m-tolylether
M-Clarity™ Qualitätsstufe = MQ100
281514-25G
281514-5G
SIAL281514-25GEA
173
EUR
SIAL281514-25G
SIAL281514-5G
5-Methyl-2-nitroanisol, Sigma-Aldrich®
5-Methyl-2-nitroanisol
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007367 CAS-Nummer: 38512-82-2 EINECS: 253-975-1 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...