2-Nitrobenzylalkohol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2-Nitrophenyl)methanol
A15496.14
A15496.22
A15496.36
A15496.14EA
49.8
EUR
A15496.14
A15496.22
A15496.36
2-Nitrobenzylalkohol ≥97%
2-Nitrobenzylalkohol
Purity: 97%
CAS No.: 612-25-9
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: 70-74°
Boiling Point: 270°
MDL No.: MFCD00007186
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Siedepunkt: 270 °C Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007186 CAS-Nummer: 612-25-9 EINECS: 210-302-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...