2-Nitrobenzylalkohol

Lieferant: Thermo Scientific

Synonyme: (2-Nitrophenyl)methanol

SB01200.1 SB01200.10 SB01200.50
MAYBSB01200.1EA 0 EUR
MAYBSB01200.1 MAYBSB01200.10 MAYBSB01200.50
2-Nitrobenzylalkohol
2-Nitrobenzylalkohol
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Siedepunkt: 270 °C (1013 hPa)
Schmelzpunkt: 71…72 °C
MDL: MFCD00007186
CAS-Nummer: 612-25-9
EINECS: 210-302-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR