3-Nitrobenzylalkohol ≥98%

Lieferant: Thermo Scientific

Synonyme: (3-Nitrophenyl)methanol

A15040.14 A15040.22
A15040.14EA 50 EUR
A15040.14 A15040.22
3-Nitrobenzylalkohol ≥98%
3-Nitrobenzylalkohol

Purity: 98% CAS No.: 619-25-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 110°(230°F) Refractive Index: 1.5755 Melting Point: 29-33° Boiling Point: 175-180°/3mm Beilstein Reference: 2044769 MDL No.: MFCD00007273

Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Siedepunkt: 175…180 °C (3 mmHg)
Schmelzpunkt: 29…33 °C
Dichte: 1,300
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007273
CAS-Nummer: 619-25-0
EINECS: 210-588-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR