4-Nitrobenzylalkohol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Nitrophenyl)methanol

A15742.14 A15742.22
A15742.14EA 33.2 EUR
A15742.14 A15742.22
4-Nitrobenzylalkohol ≥99%
4-Nitrobenzylalkohol

Purity: 99% CAS No.: 619-73-8 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 180°(356°F) Melting Point: 92-95° Boiling Point: 185°/12mm Beilstein Reference: 1424026 MDL No.: MFCD00007376

Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Siedepunkt: 185 °C (12 mmHg)
Schmelzpunkt: 92…95 °C
Flammpunkt: 180 °C (356 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007376
CAS-Nummer: 619-73-8
EINECS: 210-611-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR