3-Nitro-o-kresol

Lieferant: CHEMPUR

RH01817.250 RH01817.1
CHMPRH01817.250EA 0 EUR
CHMPRH01817.250 MAYBRH01817.1
3-Nitro-o-kresol
3-Nitro-o-kresol
Formel: C₇H₇NO₃
Schmelzpunkt: 149…152 °C
MDL: MFCD00007241
CAS-Nummer: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR