5-Nitro-o-kresol

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Hydroxy-2-methyl-5-nitrobenzol, 2-Methyl-5-nitrophenol, 2-Hydroxy-4-nitrotoluol

SB01762.1 SB01762EA SB01762EE
MAYBSB01762.1EA 0 EUR
MAYBSB01762.1 MAYBSB01762EA MAYBSB01762EE
5-Nitro-o-kresol
5-Nitro-o-kresol
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
MDL: MFCD00043909
CAS-Nummer: 5428-54-6
EINECS: 226-580-7
UN: 2446
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR