6-Nitro-m-kresol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Hydroxy-3-methyl-6-nitrobenzol, 3-Hydroxy-4-nitrotoluol, 5-Methyl-2-nitrophenol

A11729.14 A11729.22 A11729.36
A11729.14EA 34.5 EUR
A11729.14 A11729.22 A11729.36
6-Nitro-m-kresol ≥97%
6-Nitro-m-kresol

Purity: 97% CAS No.: 700-38-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 109°(228°F) Odour: Phenol-like Beilstein Reference: 1868030 UN No.: 2446 MDL No.: MFCD00007111

Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Schmelzpunkt: 53…56 °C
Flammpunkt: 109 °C (228 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007111
CAS-Nummer: 700-38-9
EINECS: 211-843-0
UN: 2446
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR