3-Methoxybenzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-(Hydroxymethyl)anisol, (3-Methoxyphenyl)methanol

A15471.14 A15471.22
A15471.14EA 41.9 EUR
A15471.14 A15471.22
3-Methoxybenzylalkohol ≥98%
3-Methoxybenzylalkohol

Purity: 98+% CAS No.: 6971-51-3 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 109°(228°F) Refractive Index: 1.5440 Boiling Point: 252-254° Beilstein Reference: 2042063 MDL No.: MFCD00004637

Formel: C₈H₁₀O₂
Molecular Weight: 138.17 g/mol
Siedepunkt: 252…254 °C
Schmelzpunkt: 29…30 °C
Dichte: 1.112
Flammpunkt: 109 °C (228 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004637
CAS-Nummer: 6971-51-3
EINECS: 230-200-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR