4-Methoxybenzylalkohol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(Hydroxymethyl)anisol, Anisalkohol, (4-Methoxyphenyl)methanol

A15559.0E A15559.22 A15559.36
A15559.0EEA 450 EUR
A15559.0E A15559.22 A15559.36
4-Methoxybenzylalkohol ≥98%
4-Methoxybenzylalkohol

Purity: 98% CAS No.: 105-13-5 Molecular Formula: C8H10O2 Formula Weight: 138.17 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Refractive Index: 1.5450 Melting Point: 22-25° Boiling Point: 259° Beilstein Reference: 636654 Merck Reference: 14,604 MDL No.: MFCD00004653

Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 259 °C
Schmelzpunkt: 22…25 °C
Dichte: 1,110
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00004653
CAS-Nummer: 105-13-5
EINECS: 203-273-6
Merck Index: 14,604

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR