2-Ethoxybenzylalkohol ≥98%
Lieferant: Thermo Scientific
Synonyme:
(2-Ethoxyphenyl)methanol
A15477.14
A15477.22
A15477.14EA
94.2
EUR
A15477.14
A15477.22
2-Ethoxybenzylalkohol ≥98%
2-Ethoxybenzylalkohol
Purity: 98%
CAS No.: 71672-75-8
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point:
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Siedepunkt: 263…265 °C Schmelzpunkt: 0 °C Dichte: 1,074 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004616 CAS-Nummer: 71672-75-8 EINECS: 275-811-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...