6-Methoxy-o-kresol
Lieferant: CHEMPUR
Synonyme:
2-Hydroxy-3-methylanisol, 1-Hydroxy-2-methoxy-6-methylbenzol, 2-Hydroxy-3-methoxytoluol, 2-Methoxy-6-methylphenol
BTB03158.250
BTB03158.1
CHMPBTB03158.250EA
0
EUR
CHMPBTB03158.250
MAYBBTB03158.1
6-Methoxy-o-kresol
6-Methoxy-o-kresol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol |
MDL:
MFCD00043771 CAS-Nummer: 2896-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...