5-Methoxy-m-kresol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzol, 3-Hydroxy-5-methoxytoluol, 3-Methoxy-5-methylphenol
H27516.06
H27516.03
H27516.06EA
131
EUR
H27516.06
H27516.03
5-Methoxy-m-kresol ≥97%
5-Methoxy-m-kresol
Purity: 97%
CAS No.: 3209-13-0
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 60-66°
MDL No.: MFCD00059261
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Schmelzpunkt: 60…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059261 CAS-Nummer: 3209-13-0 EINECS: 221-716-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...