5-Methoxy-m-kresol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzol, 3-Hydroxy-5-methoxytoluol, 3-Methoxy-5-methylphenol
008959-1G
008959-5G
FLUO008959-1GEA
60.9
EUR
FLUO008959-1G
FLUO008959-5G
5-Methoxy-m-kresol
5-Methoxy-m-kresol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol |
MDL:
MFCD00059261 CAS-Nummer: 3209-13-0 EINECS: 221-716-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...