3,5-Dimethoxybenzylalkohol ≥99%

Lieferant: Thermo Scientific

Synonyme: (3,5-Dimethoxyphenyl)methanol

B25138.06 B25138.14
B25138.06EA 68.2 EUR
B25138.06 B25138.14
3,5-Dimethoxybenzylalkohol ≥99%
3,5-Dimethoxybenzylalkohol

Purity: 99% CAS No.: 705-76-0 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Melting Point: 46-50° Boiling Point: 183-186°/20mm Beilstein Reference: 1866305 MDL No.: MFCD00004641

Formel: C₉H₁₂O₃
Molecular Weight: 168,19 g/mol
Siedepunkt: 183…186 °C (20 mmHg)
Schmelzpunkt: 46…50 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004641
CAS-Nummer: 705-76-0
EINECS: 211-888-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR