2,4,6-Trimethoxytoluol ≥97%

Lieferant: Thermo Scientific

Synonyme: 1-Methyl-2,4,6-trimethoxybenzol

L02155.06 L02155.14
L02155.06EA 86.1 EUR
L02155.06 L02155.14
2,4,6-Trimethoxytoluol ≥97%
2,4,6-Trimethoxytoluol

Purity: 97% CAS No.: 14107-97-2 Molecular Formula: C10H14O3 Formula Weight: 182.22 Refractive Index: 1.5310 Melting Point: 27-28° Boiling Point: 100-105°/1mm Beilstein Reference: 2048952 MDL No.: MFCD00008375

Formel: C₁₀H₁₄O₃
Siedepunkt: 100…105 °C (1 mmHg)
Schmelzpunkt: 27…28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008375
CAS-Nummer: 14107-97-2
EINECS: 237-962-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR