2,4,6-Trimethoxybenzylalkohol ≥97%
Lieferant: Thermo Scientific
H50348.03
H50348.MD
H50348.03EA
174
EUR
H50348.03
H50348.MD
2,4,6-Trimethoxybenzylalkohol ≥97%
2,4,6-Trimethoxybenzylalkohol
Purity: 97%
CAS No.: 61040-78-6
Molecular Formula: C10H14O4
Formula Weight: 198.22
Melting Point: 60°
MDL No.: MFCD00060356
Formel:
C₁₀H₁₄O₄ Molecular Weight: 198,22 g/mol Schmelzpunkt: 60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060356 CAS-Nummer: 61040-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...