4-Phenyl-1-butanol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Phenylbutan-1-ol

180370100 180370500
ACRO180370100EA 68.2 EUR
ACRO180370100 ACRO180370500
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol
Formel: (C₆H₅)(CH₂)₃CH₂OH
Molecular Weight: 150.22 g/mol
Siedepunkt: 140 °C (14 mmHg)
Dichte: 0.975 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00002971
CAS-Nummer: 3360-41-6
EINECS: 222-128-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20°C, 589 nm)
GC ≥96.0 %
Optical rotation +43° to +50° (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR