3-Phenoxybenzylalkohol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Hydroxymethyl)diphenylether
183770250
183771000
183770050
ACRO183770250EA
71
EUR
ACRO183770250
ACRO183771000
ACRO183770050
3-Phenoxybenzylalkohol 98%
3-Phenoxybenzylalkohol
Formel:
C₁₃H₁₂O₂ Molecular Weight: 200,24 g/mol Siedepunkt: 135…140 °C (1 mmHg) Dichte: 1,1456 g/cm³ Flammpunkt: 309 °C |
MDL:
MFCD00004636 CAS-Nummer: 13826-35-2 EINECS: 237-525-1 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5910 to 1.5950 (20°C, 589 nm) |
GC | ≥97.5 % |
Solubility | (2 % in DMSO) Clear colorless to light yellow solution |
Titration with HClO4 | 99 to 101 % (quetiapine hemifumarate dry substance) |
Titration with NaOH | 12.5 to 13.8 % (fumaric acid dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...