4-Hydroxybenzylalkohol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Hydroxyphenyl)methanol, 4-(Hydroxymethyl)phenol

A13132.18 A13132.22 A13132.30
A13132.18EA 83.8 EUR
A13132.18 A13132.22 A13132.30
4-Hydroxybenzylalkohol ≥99%
4-Hydroxybenzylalkohol

Purity: 99% CAS No.: 623-05-2 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 114-122° Boiling Point: 251-253° MDL No.: MFCD00004658

Formel: C₇H₈O₂
Molecular Weight: 124,14 g/mol
Siedepunkt: 251…253 °C
Schmelzpunkt: 114…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004658
CAS-Nummer: 623-05-2
EINECS: 210-768-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR