5-(Hydroxymethyl)resorcin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(3,5-Dihydroxyphenyl)methanol, 3,5-Dihydroxybenzylalkohol
344060050
344060250
ACRO344060050EA
75.9
EUR
ACRO344060050
ACRO344060250
5-(Hydroxymethyl)resorcin 98%
5-(Hydroxymethyl)resorcin
Formel:
C₇H₈O₃ Molecular Weight: 140.14 g/mol Schmelzpunkt: 181…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016867 CAS-Nummer: 29654-55-5 EINECS: 249-751-8 |
Spezifikation Testergebnisse
Appearance | Off-white to beige-pink to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 181 to 186 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...