5-(Hydroxymethyl)resorcin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(3,5-Dihydroxyphenyl)methanol, 3,5-Dihydroxybenzylalkohol
M-Clarity™ Qualitätsstufe = MQ100
396206-25G
396206-5G
SIAL396206-25GEA
130
EUR
SIAL396206-25G
SIAL396206-5G
5-(Hydroxymethyl)resorcin, Sigma-Aldrich®
5-(Hydroxymethyl)resorcin
Formel:
C₇H₈O₃ Molecular Weight: 140,14 g/mol |
MDL:
MFCD00016867 CAS-Nummer: 29654-55-5 EINECS: 249-751-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...