4-Toluolthiol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Thiocresol, p-Toluolthiol, 4-Methylbenzol-1-thiol, 1-Mercapto-4-methylbenzol, 4-Methylphenylmercaptan, p-Thiocresol, 4-Methylbenzolthiol, 4-Methylthiophenol
138710100
138711000
138715000
138710025
ACRO138710100EA
47.6
EUR
ACRO138710100
ACRO138711000
ACRO138715000
ACRO138710025
4-Toluolthiol 98%
4-Toluolthiol
Formel:
C₇H₈S Molecular Weight: 124.21 g/mol Siedepunkt: 195 Schmelzpunkt: 40…44 °C Flammpunkt: 68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004851 CAS-Nummer: 106-45-6 EINECS: 203-399-1 UN: 2811 ADR: 6.1 III Merck Index: 12,09461 |
Spezifikation Testergebnisse
Appearance | White to yellow Fused solid |
Infrared spectrum | Conforms |
Melting point | 40 to 44 °C |
GC | ≥97.5 % |
Elemental Analysis | 15.53 to 16.83 % N |
Specific optical rotation | +15° to +25° (20 °C, 589 nm) (c=1, water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...