Tetradecylbenzol ≥97%
Lieferant: Thermo Scientific
A19988.06
A19988.14
A19988.06EA
37
EUR
A19988.06
A19988.14
Tetradecylbenzol ≥97%
Tetradecylbenzol
Purity: 97%
CAS No.: 1459-10-5
Molecular Formula: C6H5(CH2)13CH3
Formula Weight: 274.49
Refractive Index: 1.4856
Melting Point: 9°
Boiling Point: 196°/6mm
MDL No.: MFCD00015080
Formel:
C₂₀H₃₄ Molecular Weight: 274,49 g/mol Siedepunkt: 196 °C (6 mmHg) Schmelzpunkt: 9 °C Dichte: 0,8565 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015080 CAS-Nummer: 1459-10-5 EINECS: 215-950-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...