1-tert-Butyl-4-ethinylbenzol 96%

Lieferant: Thermo Fisher Scientific
Achtung

380850050 380850250
ACRO380850050EA 56.1 EUR
ACRO380850050 ACRO380850250
1-tert-Butyl-4-ethinylbenzol 96%
1-tert-Butyl-4-ethinylbenzol
Formel: C₁₂H₁₄
Siedepunkt: 130 °C (40 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190197
CAS-Nummer: 772-38-3
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR