1-tert-Butyl-4-ethinylbenzol
Lieferant: FLUOROCHEM
Synonyme:
1-Ethinyl-4-tert-butylbenzol, (4-tert-Butylphenyl)acetylen, 4-Ethinyl-1-tert-butylbenzol, (4-tert-Butylphenyl)ethin, 4-tert-Butyl-1-ethinylbenzol
CFF875.
009071-10G
009071-50G
CHMPCFF875.EA
0
EUR
CHMPCFF875.
FLUO009071-10G
FLUO009071-50G
1-tert-Butyl-4-ethinylbenzol
1-tert-Butyl-4-ethinylbenzol
Formel:
C₁₂H₁₄ Molecular Weight: 158,24 g/mol |
MDL:
MFCD00190197 CAS-Nummer: 772-38-3 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...