1-tert-Butyl-4-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethinyl-4-tert-butylbenzol, (4-tert-Butylphenyl)acetylen, 4-Ethinyl-1-tert-butylbenzol, (4-tert-Butylphenyl)ethin, 4-tert-Butyl-1-ethinylbenzol

M-Clarity™ Qualitätsstufe = MQ100

593001-25G 593001-5G
SIAL593001-25GEA 241 EUR
SIAL593001-25G SIAL593001-5G
1-tert-Butyl-4-ethinylbenzol, Sigma-Aldrich®
1-tert-Butyl-4-ethinylbenzol
Formel: C₁₂H₁₄
Molecular Weight: 158.24 g/mol
MDL: MFCD00190197
CAS-Nummer: 772-38-3
UN: 3082
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR