1-Pentyl-4-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Pentyl-1-ethinylbenzol, (4-Pentylphenyl)acetylen, 1-Ethinyl-4-pentylbenzol, 4-Ethinyl-1-pentylbenzol, (4-Pentylphenyl)ethin

M-Clarity™ Qualitätsstufe = MQ100

457477-10G 457477-1G
SIAL457477-10GEA 101 EUR
SIAL457477-10G SIAL457477-1G
1-Pentyl-4-ethinylbenzol, Sigma-Aldrich®
1-Pentyl-4-ethinylbenzol
Formel: C₁₃H₁₆
Molecular Weight: 172,27 g/mol
MDL: MFCD00173884
CAS-Nummer: 79887-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR