Bromthymolblau ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
3′,3′′-Dibromthymolsulfonphthalein, BTB
32638.06
32638.14
32638.06EA
38.5
EUR
32638.06
32638.14
Bromthymolblau ACS
Bromthymolblau
CAS No.: 76-59-5
Molecular Formula: C27H28Br2O5S
Formula Weight: 624.39
Physical Form: Crystalline
Appearance: Pale brown to red
Melting Point: 200-202°
Beilstein Reference: 373934
Merck Reference: 14,1445
MDL No.: MFCD00005872
Formel:
C₂₇H₂₈Br₂O₅S Molecular Weight: 624.39 g/mol Schmelzpunkt: 200…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005872 CAS-Nummer: 76-59-5 EINECS: 200-971-2 Merck Index: 14,1445 |