3-Chlorbenzophenon ≥98%

Lieferant: Thermo Scientific

Synonyme: m-Chlorobenzophenone

B23085.06 B23085.14 B23085.22
B23085.06EA 31.9 EUR
B23085.06 B23085.14 B23085.22
3-Chlorbenzophenon ≥98%
3-Chlorbenzophenon

Purity: 98+% CAS No.: 1016-78-0 Molecular Formula: C13H9ClO Formula Weight: 216.67 Melting Point: 81-85° Beilstein Reference: 1869822

Formel: C₁₃H₉ClO
Molecular Weight: 216,67 g/mol
Siedepunkt: 322 °C (1013 hPa)
Schmelzpunkt: 81…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009816
CAS-Nummer: 1016-78-0
EINECS: 213-809-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR