4-Aminobenzophenon 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Benzoylanilin, p-Aminobenzophenon
A13757.09
A13757.18
A13757.09EA
53
EUR
A13757.09
A13757.18
4-Aminobenzophenon 98%
4-Aminobenzophenon
Purity: 98%
CAS No.: 1137-41-3
Molecular Formula: C13H11NO
Formula Weight: 197.24
Melting Point: 121-124°
Boiling Point: 245-247°/22mm
MDL No.: MFCD00007895
Formel:
C₁₃H₁₁NO Molecular Weight: 197,24 g/mol Siedepunkt: 245…247 °C (22 mmHg) Schmelzpunkt: 121…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007895 CAS-Nummer: 1137-41-3 EINECS: 214-506-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...