2-Methoxybenzophenon ≥98%
Lieferant: Thermo Scientific
B23376.03
B23376.06
B23376.03EA
54
EUR
B23376.03
B23376.06
2-Methoxybenzophenon ≥98%
2-Methoxybenzophenon
Purity: 98%
CAS No.: 2553-04-0
Molecular Formula: C14H12O2
Formula Weight: 212.25
Melting Point: 36-38°
Boiling Point: 149-150°/1mm
MDL No.: MFCD00017163
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 149…150 °C (1 mmHg) Schmelzpunkt: 36…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017163 CAS-Nummer: 2553-04-0 EINECS: 219-857-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...