4-Hydroxybenzophenon 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
p-Hydroxybenzophenon
121010250
121011000
ACRO121010250EA
32.2
EUR
ACRO121010250
ACRO121011000
4-Hydroxybenzophenon 98+%
4-Hydroxybenzophenon
Formel:
C₁₃H₁₀O₂ Molecular Weight: 198,22 g/mol Siedepunkt: 367 °C (1013 hPa) Schmelzpunkt: 132…135 °C Dichte: 1,133 g/cm³ (172 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002355 CAS-Nummer: 1137-42-4 EINECS: 214-507-1 |
Spezifikation Testergebnisse
Appearance | White to beige to brown Powder |
Infrared spectrum | Conforms |
Melting point | 132 to 135 °C |
GC | ≥98.0 % |
Sodium chloride (NaCl) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤0.5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...