2-Fluorbenzophenon

Lieferant: FLUOROCHEM
Achtung

Synonyme: o-Fluorobenzophenone

003865-10G 003865-25G 003865-100G
FLUO003865-10GEA 0 EUR
FLUO003865-10G FLUO003865-25G FLUO003865-100G
2-Fluorbenzophenon
2-Fluorbenzophenon
Formel: C₁₃H₉FO
Molecular Weight: 200.21 g/mol
Siedepunkt: 95…96 °C (0.05 mmHg)
Dichte: 1.18 g/cm³ (20 °C)
MDL: MFCD00000318
CAS-Nummer: 342-24-5
EINECS: 206-440-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR