4-Fluorbenzophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11185.14
A11185.22
A11185.36
A11185.14EA
45.3
EUR
A11185.14
A11185.22
A11185.36
4-Fluorbenzophenon ≥97%
4-Fluorbenzophenon
Purity: 97%
CAS No.: 345-83-5
Molecular Formula: C13H9FO
Formula Weight: 200.21
Flash Point:
Formel:
C₁₃H₉FO Molecular Weight: 200.21 g/mol Siedepunkt: 130…132 °C (3 mmHg) Schmelzpunkt: 45…49 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000352 CAS-Nummer: 345-83-5 EINECS: 206-463-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...